Learn More
Isoprocarb in Methanol 10μg/mL, Fisher Chemical™
Product Code. 12967043 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
10mL
Product Code. Quantity unitSize
12967043 1 mL 10mL
1 options
Product Code. 12967043
Brand: Fisher Chemical P880120
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Packaging Ampoule
CAS 99675-03-3
Analyte or Component Names Isofenphos-methyl
Molecular Formula C14H22NO4PS
Concentration 100 μg/mL
InChI Key IXTOWLKEARFCCP-UHFFFAOYSA-N
SMILES CC(C)NP(=S)(OC)OC1=CC=CC=C1C(=O)OC(C)C
Element Isofenphos-methyl
IUPAC Name propan-2-yl 2-[methoxy-(propan-2-ylamino)phosphinothioyl]oxybenzoate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 331.367
PubChem CID 127394
Quantity 1 mL
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Standard solution Isoprocarb,2-isopropylphenyl N-methylcarbamate 10ug/ml in methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P501-Dispose of contents/container to .