accessibility menu, dialog, popup

UserName

CAS RN 99067-15-9

IUPAC Name: 4-bromo-6-(2,3-dihydro-1H-pyrazol-3-ylidene)cyclohexa-2,4-dien-1-one
Synonyms: 4-bromo-6-(1,2-dihydropyrazol-3-ylidene)cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 239.07
Molecular Formula: C9H7BrN2O
InChi Key: WAHULEHSSFNFQX-UHFFFAOYSA-N
SMILES: BrC1=CC(=C2NNC=C2)C(=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results