CAS RN 97627-24-2
CAS RN 97627-24-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(2-Chloro-6-methylphenyl)-N'-4-pyridinylurea, MedChemExpress
MedChemExpress N-[(1R)-4-[(Aminoiminomethyl)amino]-1-[[[(1R)-1-(4-hydroxyphenyl)ethyl]amino]carbonyl]butyl]-α-phenylbenzeneacetamide is an anticonvulsant agent with potential for the treatment of generalized tonic-clonic and partial seizures.