Learn More
N-(2-Chloro-6-methylphenyl)-N'-4-pyridinylurea, MedChemExpress
Product Code. 16463518
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463518 10 mg 10mg
16440869 1 mg 1mg
16430496 20 mg 20mg
16416247 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16463518 Supplier Medchem Express Supplier No. HY101708
MedChemExpress N-[(1R)-4-[(Aminoiminomethyl)amino]-1-[[[(1R)-1-(4-hydroxyphenyl)ethyl]amino]carbonyl]butyl]-α-phenylbenzeneacetamide is an anticonvulsant agent with potential for the treatment of generalized tonic-clonic and partial seizures.
N-[(1R)-4-[(Aminoiminomethyl)amino]-1-[[[(1R)-1-(4-hydroxyphenyl)ethyl]amino]carbonyl]butyl]-α-phenylbenzeneacetamide is an anticonvulsant agent with potential for the treatment of generalized tonic-clonic and partial seizures.
Specifications
Chemical Name or Material N-(2-Chloro-6-methylphenyl)-N'-4-pyridinylurea
CAS 97627-24-2
Color White
Purity Grade Notes Research
Molecular Formula C13H12ClN3O
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (477.63 mM; Need ultrasonic)
SMILES O=C(NC1=CC=NC=C1)NC2=C(C)C=CC=C2Cl
Molecular Weight (g/mol) 261.71
Formula Weight 261.71
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation