CAS RN 95975-55-6
CAS RN 95975-55-6
IUPAC Name:
(1E,3aS,3bR,9aR,9bS,11aS)-1-ethylidene-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-2,7-dione
Synonyms:
guggulsterone
Molecular Weight (g/mol):
312.45
Molecular Formula:
C21H28O2
InChi Key:
WDXRGPWQVHZTQJ-AUKWTSKRSA-N
SMILES:
C\C=C1\C(=O)C[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
1
–
1
of
1
results
1
Guggulsterone, MedChemExpress
MedChemExpress Guggulsterone is a plant sterol derived from the gum resin of the tree Commiphora wightii.