Learn More
Z/E-Guggulsterone, TRC
Specifications
Specifications
| Chemical Name or Material | Z/E-Guggulsterone |
| CAS | 95975-55-6 |
| Molecular Formula | C21H28O2 |
| Quantity | 10 mg |
| Synonym | Guggulsterone, Guglip, Gugulipid |
| InChI Formula | InChI=1S/C21H28O2/c1-4-16-19(23)12-18-15-6-5-13-11-14(22)7-9-20(13,2)17(15)8-10-21(16,18)3/h4,11,15,17-18H,5-10,12H2,1-3H3/t15-,17+,18+,20+,21-/m1/s1 |
| SMILES | C\C=C\1/C(=O)C[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | (8R,9S,10R,13S,14S)-17-ethylidene-10,13-dimethyl-1,2,6,7,8,9,11,12,14,15-decahydrocyclopenta[a]phenanthrene-3,16-dione |
| Molecular Weight (g/mol) | 312.45 |
| Formula Weight | 312.2089 |