accessibility menu, dialog, popup

UserName

CAS RN 95883-10-6

IUPAC Name: 3-(2,5-dimethylphenyl)prop-2-enoic acid
Synonyms: 3-(2,5-dimethylphenyl)prop-2-enoic acid
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: FAIBVLSPNAHVSQ-UHFFFAOYSA-N
SMILES: CC1=CC(C=CC(O)=O)=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results