accessibility menu, dialog, popup

UserName

HOOHOHONBrOHNO

CAS RN 957-75-5

IUPAC Name: 5-bromo-1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 5-bromouridine
Molecular Weight (g/mol): 323.1
Molecular Formula: C9H11BrN2O6
InChi Key: AGFIRQJZCNVMCW-UHFFFAOYNA-N
SMILES: OCC1OC(C(O)C1O)N1C=C(Br)C(=O)NC1=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)