Learn More
5-Bromouridine, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Bromouridine |
| CAS | 957-75-5 |
| Molecular Formula | C9H11BrN2O6 |
| Quantity | 25 g |
| Synonym | (-)-5-Bromouridine, NSC 38296 |
| InChI Formula | InChI=1S/C9H11BrN2O6/c10-3-1-12(9(17)11-7(3)16)8-6(15)5(14)4(2-13)18-8/h1,4-6,8,13-15H,2H2,(H,11,16,17)/t4-,5-,6-,8-/m1/s1 |
| SMILES | OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=C(Br)C(=O)NC2=O |
| IUPAC Name | 5-bromo-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione |
| Molecular Weight (g/mol) | 323.1 |
| Formula Weight | 321.98 |