CAS RN 956274-94-5
CAS RN 956274-94-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Mavatrep, MedChemExpress
MedChemExpress Mavatrep is an orally bioavailable TRPV1 antagonist (Ki=6.5 nM), exhibits minimal effect on the enzymatic activity (IC50 > 25 μM) of CYP isoforms 3A4, 1A2, and 2D6.