Learn More
Mavatrep, MedChemExpress
Product Code. 16437218
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16437218
Brand: Medchem Express HY16935
MedChemExpress Mavatrep is an orally bioavailable TRPV1 antagonist (Ki=6.5 nM), exhibits minimal effect on the enzymatic activity (IC50 > 25 μM) of CYP isoforms 3A4, 1A2, and 2D6.
Mavatrep is an orally bioavailable TRPV1 antagonist (Ki=6.5 nM), exhibits minimal effect on the enzymatic activity (IC50 > 25 μM) of CYP isoforms 3A4, 1A2, and 2D6.
Specifications
Chemical Name or Material Mavatrep
Physical Form Solid
CAS 956274-94-5
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C25H21F3N2O
Synonym JNJ-39439335
Solubility Information DMSO : 16.67 mg/mL (39.46 mM; ultrasonic and warming and heat to 60°C)
SMILES OC(C)(C)C1=CC=CC=C1C2=CC=C3N=C(/C=C/C4=CC=C(C(F)(F)F)C=C4)NC3=C2
Molecular Weight (g/mol) 422.44
Formula Weight 422.44
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.