accessibility menu, dialog, popup

UserName

CAS RN 948006-26-6

IUPAC Name: 3-(ethoxycarbonyl)-2-hydroxy-5,6,7,8-tetrahydronaphthalene-1-carboxylic acid
Synonyms: 3-(ethoxycarbonyl)-2-hydroxy-5,6,7,8-tetrahydronaphthalene-1-carboxylic acid
Molecular Weight (g/mol): 264.28
Molecular Formula: C14H16O5
InChi Key: NQDPXKMLMJUBIJ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=C(CCCC2)C(C(O)=O)=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results