accessibility menu, dialog, popup

UserName

CAS RN 94724-12-6

H3CNHOH3COH3CNHCH3OCH3HNCH3HClHClHCl

CAS RN 94724-12-6

IUPAC Name: [2-(4-methoxy-3-{[2-methoxy-3-({2-methoxy-5-[2-(methylamino)ethyl]phenyl}methyl)-5-[2-(methylamino)ethyl]phenyl]methyl}phenyl)ethyl](methyl)amine trihydrochloride
Synonyms: [2-(4-methoxy-3-{[2-methoxy-3-({2-methoxy-5-[2-(methylamino)ethyl]phenyl}methyl)-5-[2-(methylamino)ethyl]phenyl]methyl}phenyl)ethyl](methyl)amine trihydrochloride
Molecular Weight (g/mol): 629.1
Molecular Formula: C32H48Cl3N3O3
InChi Key: KIAWAQJORBNOIF-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.CNCCC1=CC(CC2=CC(CCNC)=CC(CC3=C(OC)C=CC(CCNC)=C3)=C2OC)=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results