Learn More
Compound 48/80, MP Biomedicals™
Product Code. 11474322 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474322 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11474322 Supplier MP Biomedicals Supplier No. 0215902650
  • ONLINE EXCLUSIVE PRICE
    2280.00 NOK
    2167.40 NOK / 50mg
    Save 112.60 NOK
    4% Off
 Restricted Item
Estimated Shipment: 19-08-2026
Specifications
CAS 94724-12-6
Color White
Molecular Formula C32H48Cl3N3O3
Quantity 50 mg
Synonym compound 48/80 trihydrochloride
InChI Key KIAWAQJORBNOIF-UHFFFAOYSA-N
SMILES CNCCC1=CC(=C(C=C1)OC)CC2=CC(=CC(=C2OC)CC3=C(C=CC(=C3)CCNC)OC)CCNC.Cl.Cl.Cl
IUPAC Name 2-[4-methoxy-3-[[2-methoxy-3-[[2-methoxy-5-[2-(methylamino)ethyl]phenyl]methyl]-5-[2-(methylamino)ethyl]phenyl]methyl]phenyl]-N-methylethanamine;trihydrochloride
Molecular Weight (g/mol) 629.104
PubChem CID 118797323
Formula Weight 630
Physical Form Powder
Chemical Name or Material Compound 48/80
Show More Show Less
compliance-icons
Product Identifier
  • Compound 48/80
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Compound 48/80