CAS RN 93951-69-0
CAS RN 93951-69-0
IUPAC Name:
(²H₁₀)fluoranthene
Synonyms:
(²H₁₀)fluoranthene
Molecular Weight (g/mol):
212.32
Molecular Formula:
C16H10
InChi Key:
GVEPBJHOBDJJJI-LHNTUAQVSA-N
SMILES:
[2H]C1=C([2H])C([2H])=C2C(=C1[2H])C1=C([2H])C([2H])=C([2H])C3=C([2H])C([2H])=C([2H])C2=C13
1
–
7
of
7
results
2
Fluoranthene-d10, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Fluoranthene-d10, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Fluoranthene-d10, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
5
Fluoranthene-d10, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.