Learn More
Fluoranthene-d10, TRC
Specifications
Specifications
| Chemical Name or Material | Fluoranthene D10 |
| CAS | 93951-69-0 |
| Molecular Formula | C16 2H10 |
| Quantity | 100 mg |
| Synonym | Fluoranthene-1,2,3,4,5,6,7,8,9,10-d10, Fluoranthene-d10 (9CI), Perdeuterated fluoranthene, [10-2H]Fluoranthene, Fluoranthene D10 |
| InChI Formula | InChI=1S/C16H10/c1-2-8-13-12(7-1)14-9-3-5-11-6-4-10-15(13)16(11)14/h1-10H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D |
| SMILES | [2H]c1c([2H])c([2H])c2c(c1[2H])c3c([2H])c([2H])c([2H])c4c([2H])c([2H])c([2H])c2c34 |
| IUPAC Name | 1,2,3,4,5,6,7,8,9,10-decadeuteriofluoranthene |
| Molecular Weight (g/mol) | 212.31 |
| Formula Weight | 212.141 |