accessibility menu, dialog, popup

UserName

BNFClHOOH

CAS RN 937595-70-5

IUPAC Name: (5-chloro-2-fluoropyridin-3-yl)boronic acid
Synonyms: 5-chloro-2-fluoropyridin-3-ylboronic acid
Molecular Weight (g/mol): 175.35
Molecular Formula: C5H4BClFNO2
InChi Key: ZUDXVHDVUTUTFH-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(Cl)=CN=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results