CAS RN 932372-01-5
CAS RN 932372-01-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Asapiprant, MedChemExpress
MedChemExpress Asapiprant is a potent and selective DP1 receptor antagonist with a Ki of 0.44 nM.
