Learn More
Asapiprant, MedChemExpress
Product Code. 16463207
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. 16463207
Brand: Medchem Express HY16763
MedChemExpress Asapiprant is a potent and selective DP1 receptor antagonist with a Ki of 0.44 nM.
Asapiprant is a potent and selective DP1 receptor antagonist with a Ki of 0.44 nM.
Specifications
Chemical Name or Material Asapiprant
CAS 932372-01-5
Color White
Purity Grade Notes Research
Molecular Formula C24H27N3O7S
Quantity 50 mg
Synonym S-555739
Solubility Information DMSO : 11.11 mg/mL (22.15 mM; Need ultrasonic)
SMILES O=C(O)COC1=CC(N2CCN(S(=O)(C3=CC=C(OC(C)C)C=C3)=O)CC2)=CC=C1C4=NC=CO4
Molecular Weight (g/mol) 501.55
Formula Weight 501.55
Percent Purity 99.4%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.