accessibility menu, dialog, popup

UserName

CAS RN 920-39-8

CH3CH3MgBr

CAS RN 920-39-8

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 147.3
Molecular Formula: C3H7BrMg
InChi Key: LVKCSZQWLOVUGB-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].C[CH-]C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)