Learn More
Isopropylmagnesium bromide, 3M in 2-MeTHF
Product Code. 15481008
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. Quantity unitSize
15481008 100 mL 100mL
1 options
Product Code. 15481008
Brand: Thermo Scientific Alfa Aesar H54840.AE
Request Bulk or Custom Format
CAS: 920-39-8 | C3H7BrMg | 147.30 g/mol
Isopropylmagnesium bromide is a Grignard reagent used in organic synthesis, especially for the introduction of iso-propyl group to carbonyl compounds. It is also used as a strong base in aprotic solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isopropylmagnesium bromide is a Grignard reagent used in organic synthesis, especially for the introduction of iso-propyl group to carbonyl compounds. It is also used as a strong base in aprotic solvent.
Solubility
TRUSTED_SUSTAINABILITY
Specifications
CAS 920-39-8
Molecular Formula C3H7BrMg
Linear Formula (CH3)2CHMgBr
MDL Number MFCD00672110
Quantity 100 mL
UN Number UN3399
Sensitivity Air and moisture sensitive; May form a precipitate
Synonym isopropylmagnesium bromide, magnesium, bromo 1-methylethyl, isopropylmagnesiumbromide, bromo 1-methylethyl magnesium, iprmgbr, i-prmgbr, bromoisopropylmagnesium, i-propylmagnesiumbromide, i-propylmagnesium bromide, bromo-isopropyl-magnesium, grignard reagent
Solubility Information Miscible with tetrahydorfuran and diethyl ether. Immiscible with hydrocarbons.
InChI Key UCJIVFREPMUZDC-UHFFFAOYSA-M
SMILES CC(C)[Mg]Br
IUPAC Name magnesium;propane;bromide
Molecular Weight (g/mol) 147.30
PubChem CID 101909
Formula Weight 147.3
Concentration or Composition (by Analyte or Components) 3M in 2-MeTHF
Chemical Name or Material Isopropylmagnesium bromide