CAS RN 912999-49-6
CAS RN 912999-49-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Onalespib, MedChemExpress
MedChemExpress Onalespib (AT13387) is a long-acting second-generation Hsp90 inhibitor with a Kd of 0.71 nM.