Learn More
Onalespib, MedChemExpress
Product Code. 16498677
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16498677 5 mg 5mg
16466078 10 mg 10mg
16456476 100 mg 100mg
16452217 50 mg 50mg
4 options
Product Code. 16498677
Brand: Medchem Express HY14463
MedChemExpress Onalespib (AT13387) is a long-acting second-generation Hsp90 inhibitor with a Kd of 0.71 nM.
Onalespib (AT13387) is a long-acting second-generation Hsp90 inhibitor with a Kd of 0.71 nM.
Specifications
Chemical Name or Material Onalespib
CAS 912999-49-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H31N3O3
Quantity 5 mg
Synonym AT13387
Solubility Information DMSO : 50 mg/mL (122.09 mM; ultrasonic and warming and heat to 60°C)
SMILES OC1=CC(O)=C(C(C)C)C=C1C(N2CC(C=CC(CN3CCN(C)CC3)=C4)=C4C2)=O
Molecular Weight (g/mol) 409.52
Formula Weight 409.52
Percent Purity 98.44%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death