accessibility menu, dialog, popup

UserName

CAS RN 910037-04-6

H3CNHONCH3CH3

CAS RN 910037-04-6

IUPAC Name: dimethyl(3-{4-[(methylamino)methyl]phenoxy}propyl)amine
Synonyms: dimethyl(3-{4-[(methylamino)methyl]phenoxy}propyl)amine
Molecular Weight (g/mol): 222.33
Molecular Formula: C13H22N2O
InChi Key: LFLFEMOINFHORX-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(OCCCN(C)C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results