CAS RN 91-94-1
CAS RN 91-94-1
IUPAC Name:
3,3'-dichloro-[1,1'-biphenyl]-4,4'-diamine
Synonyms:
3,3'-dichlorobenzidine
Molecular Weight (g/mol):
253.13
Molecular Formula:
C12H10Cl2N2
InChi Key:
HUWXDEQWWKGHRV-UHFFFAOYSA-N
SMILES:
NC1=CC=C(C=C1Cl)C1=CC=C(N)C(Cl)=C1
1
–
5
of
5
results
1
Benzidines Mixture 1 2000 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
3
EPA Method 8270 Benzidines Mixture 2000 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality