Learn More
EPA Method 8270 Benzidines Mixture 2000 μg/mL in Methanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | 3,3'-Dichlorobenzidine∣3,3'-Dimethylbenzidine∣4,4'-Benzidine |
| CAS | 91-94-1, 119-93-7, 92-87-5 |
| Analyte or Component Names | 3,3'-Dichlorobenzidine∣3,3'-Dimethylbenzidine∣4,4'-Benzidine |
| Shipping Condition | Ice pack (-20°C) |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 3,3'-Dichlorobenzidine 2000 μg/mL∣3,3'-Dimethylbenzidine 2000 μg/mL∣4,4'-Benzidine 2000 μg/mL |