CAS RN 905854-02-6
CAS RN 905854-02-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tivantinib, MedChemExpress
MedChemExpress Tivantinib is a highly selective c-Met tyrosine kinase inhibitor with a Ki of 355 nM.

2