Learn More
Tivantinib, TRC
Specifications
Specifications
| Chemical Name or Material | Tivantinib |
| CAS | 905854-02-6 |
| Molecular Formula | C23H19N3O2 |
| Quantity | 5 mg |
| Synonym | (3R,4R)-3-(5,6-Dihydro-4H-pyrrolo[3,2,1-ij]quinolin-1-yl)-4-(1H-indol-3-yl)pyrrolidine-2,5-dione, ARQ 197 |
| InChI Formula | InChI=1S/C23H19N3O2/c27-22-19(16-11-24-18-9-2-1-7-14(16)18)20(23(28)25-22)17-12-26-10-4-6-13-5-3-8-15(17)21(13)26/h1-3,5,7-9,11-12,19-20,24H,4,6,10H2,(H,25,27,28)/t19-,20-/m0/s1 |
| SMILES | O=C1NC(=O)[C@H]([C@@H]1c2c[nH]c3ccccc23)c4cn5CCCc6cccc4c56 |
| Molecular Weight (g/mol) | 369.42 |
| Formula Weight | 369.1477 |