accessibility menu, dialog, popup

UserName

CAS RN 897940-14-6

FFFFOOH

CAS RN 897940-14-6

IUPAC Name: 2-[3-fluoro-2-(trifluoromethyl)phenyl]acetic acid
Synonyms: [3-fluoro-2-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol): 222.14
Molecular Formula: C9H6F4O2
InChi Key: MQDCRGGUZQIVBB-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C(C(F)=CC=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results