accessibility menu, dialog, popup

UserName

CH3FNCl

CAS RN 886500-98-7

IUPAC Name: 2-chloro-6-fluoro-3-methylbenzonitrile
Synonyms: 2-chloro-6-fluoro-3-methylbenzonitrile
Molecular Weight (g/mol): 169.58
Molecular Formula: C8H5ClFN
InChi Key: UHLLSLUQSGFENQ-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C(C#N)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results