accessibility menu, dialog, popup

UserName

BrFFFO

CAS RN 861928-27-0

IUPAC Name: 4-bromo-2-(trifluoromethyl)benzaldehyde
Synonyms: 4-bromo-2-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 253.02
Molecular Formula: C8H4BrF3O
InChi Key: PROPHDFGNNBHJX-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(C=O)C=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results