CAS RN 775351-88-7
CAS RN 775351-88-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Corylifol A, MedChemExpress
MedChemExpress Corylifol A inhibits IL-6-induced STAT3 activation and phosphorylation, with an IC50 of 0.81 μM.