accessibility menu, dialog, popup

UserName

H2NONHOHO

CAS RN 7377-08-4

IUPAC Name: 3-[(4-aminophenyl)formamido]propanoate
Synonyms: 3-[(4-aminophenyl)formamido]propanoate
Molecular Weight (g/mol): 207.21
Molecular Formula: C10H11N2O3
InChi Key: VHAXWROFYVPXMZ-UHFFFAOYSA-M
SMILES: NC1=CC=C(C=C1)C(=O)NCCC([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)