accessibility menu, dialog, popup

UserName

NClClFF

CAS RN 71701-93-4

IUPAC Name: 2,5-dichloro-3-(difluoromethyl)pyridine
Synonyms: 2,5-dichloro-3-(difluoromethyl)pyridine
Molecular Weight (g/mol): 197.99
Molecular Formula: C6H3Cl2F2N
InChi Key: GDFCUTHAIBYLSX-UHFFFAOYSA-N
SMILES: FC(F)C1=CC(Cl)=CN=C1Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results