CAS RN 71653-64-0
IUPAC Name:
2-(difluoromethoxy)benzaldehyde
Synonyms:
2-(difluoromethoxy)benzaldehyde
Molecular Weight (g/mol):
172.13
Molecular Formula:
C8H6F2O2
InChi Key:
QPBNHDFPMRENBC-UHFFFAOYSA-N
SMILES:
FC(F)OC1=CC=CC=C1C=O
1
–
1
of
1
results
1
2-(Difluoromethoxy)benzaldehyde, 97%, Thermo Scientific™
CAS: 71653-64-0 Molecular Formula: C8H6F2O2 Molecular Weight (g/mol): 172.131 MDL Number: MFCD00042251 InChI Key: QPBNHDFPMRENBC-UHFFFAOYSA-N Synonym: 2-difluoromethoxy benzaldehyde,2-difluoromethoxy-benzaldehyde,o-difluoromethoxybenzaldehyde,2-difloromethoxybenzaldehyde,pubchem2906,acmc-209ojk,difluoromethoxybenzaldehyde,ksc356m9l,zerenex zx009675,benzaldehyde, difluoromethoxy PubChem CID: 2736984 IUPAC Name: 2-(difluoromethoxy)benzaldehyde SMILES: C1=CC=C(C(=C1)C=O)OC(F)F