accessibility menu, dialog, popup

UserName

CAS RN 7149-70-4

IUPAC Name: 1-bromo-2-methyl-4-nitrobenzene
Synonyms: 1-bromo-2-methyl-4-nitrobenzene
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: HIMGPQVBNICCGL-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1Br)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results