accessibility menu, dialog, popup

UserName

H3CCH3SiClOSiClCH3CH3CH3H3CCH3H3C

CAS RN 69304-37-6

IUPAC Name: chloro({[chlorobis(propan-2-yl)silyl]oxy})bis(propan-2-yl)silane
Synonyms: chloro[(chlorodiisopropylsilyl)oxy]diisopropylsilane
Molecular Weight (g/mol): 315.43
Molecular Formula: C12H28Cl2OSi2
InChi Key: DDYAZDRFUVZBMM-UHFFFAOYSA-N
SMILES: CC(C)[Si](Cl)(O[Si](Cl)(C(C)C)C(C)C)C(C)C
Molecular Weight (g/mol) 
  • (1)
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)