CAS RN 66611-37-8
CAS RN 66611-37-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
BGP-15, MedChemExpress
MedChemExpress BGP-15 is a PARP inhibitor, with an IC50 and a Ki of 120 and 57 μM, respectively.