Learn More
BGP-15, MedChemExpress
Product Code. 16462656
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Product Code. 16462656
Brand: Medchem Express HY100828
MedChemExpress BGP-15 is a PARP inhibitor, with an IC50 and a Ki of 120 and 57 μM, respectively.
BGP-15 is a PARP inhibitor, with an IC50 and a Ki of 120 and 57 μM, respectively.
Specifications
Chemical Name or Material BGP-15
Physical Form Solid
CAS 66611-37-8
Color Light Yellow
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C14H24Cl2N4O2
For Use With (Application) COVID-19-anti-virus
Solubility Information H2O : 100 mg/mL (284.68 mM; Need ultrasonic) ∣DMSO : 11.33 mg/mL (32.25 mM; Need ultrasonic and warming)
SMILES N=C(C1=CC=CN=C1)NOCC(O)CN2CCCCC2.[H]Cl.[H]Cl
Molecular Weight (g/mol) 351.27
Formula Weight 351.27
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.