CAS RN 64013-70-3
CAS RN 64013-70-3
IUPAC Name:
2-(aminomethyl)-6-[(4,6-diamino-3-{[4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2-hydroxycyclohexyl)oxy]oxane-3,4,5-triol; bis(sulfuric acid)
Synonyms:
kanamycin; bis(sulfuric acid)
Molecular Weight (g/mol):
680.65
Molecular Formula:
C18H40N4O19S2
InChi Key:
OGTKIXVMLDAMNU-UHFFFAOYNA-N
SMILES:
OS(O)(=O)=O.OS(O)(=O)=O.NCC1OC(OC2C(N)CC(N)C(OC3OC(CO)C(O)C(N)C3O)C2O)C(O)C(O)C1O
1
–
2
of
2
results
1
Kanamycin disulfate, 700μg/mg, Thermo Scientific™
CAS: 64013-70-3 Molecular Formula: C18H40N4O19S2 Molecular Weight (g/mol): 680.647 MDL Number: MFCD00143916 InChI Key: OGTKIXVMLDAMNU-KNQICTBBSA-N Synonym: Kanamycin A; Kanamycin acid sulfate PubChem CID: 24721551 IUPAC Name: (2R,3S,4S,5R,6R)-2-(aminomethyl)-6-[(1R,2R,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid SMILES: C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O.OS(=O)(=O)O
2
TOKU-E™ Kanamycin Acid Sulfate ReadyMade™ Solution
Kanamycin Acid Sulfate ReadyMade™ Solution is provided as a sterile-filtered solution of Kanamycin Acid Sulfate in water.