CAS RN 640-61-9
CAS RN 640-61-9
IUPAC Name:
N,4-dimethylbenzene-1-sulfonamide
Synonyms:
N,4-dimethylbenzenesulfonamide
Molecular Weight (g/mol):
185.24
Molecular Formula:
C8H11NO2S
InChi Key:
GWLOGZRVYXAHRE-UHFFFAOYSA-N
SMILES:
CNS(=O)(=O)C1=CC=C(C)C=C1
1
–
1
of
1
results
1
N-Methyl-p-toluenesulfonamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.