Learn More
N-Methyl-p-toluenesulfonamide, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
N-Methyl-p-toluenesulfonamide
Specifications
Specifications
| Chemical Name or Material | N-Methyl-p-toluenesulfonamide |
| CAS | 640-61-9 |
| Analyte or Component Names | N-Methyl-p-toluenesulfonamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H11 N O2 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzenesulfonamide, N,4-dimethyl-, p-Toluenesulfonamide, N-methyl- (6CI,7CI,8CI), N,4-Dimethylbenzenesulfonamide, (p-Tolylsulfonyl)methylamine, 4,N-Dimethylbenzenesulfonamide, 4-Methyl-N-methylbenzenesulfonamide, N-Methyl-4-methylbenzenesulfonamide, N-Methyl-p-toluenesulphonamide, N-Methyl-para-toluenesulfonamide, NSC 2223, NSC 36991 |
| InChI Formula | InChI=1S/C8H11NO2S/c1-7-3-5-8(6-4-7)12(10,11)9-2/h3-6,9H,1-2H3 |
| SMILES | CNS(=O)(=O)c1ccc(C)cc1 |
| IUPAC Name | N,4-dimethylbenzenesulfonamide |
| Show More |