CAS RN 638132-34-0
CAS RN 638132-34-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
ONO-7300243, MedChemExpress
MedChemExpress ONO-7300243 is a novel, potent lysophosphatidic acid receptor 1 (LPA1) antagonist with IC50 of 0.16 μM.