CAS RN 6342-17-2
CAS RN 6342-17-2
IUPAC Name:
1-[4-(prop-2-enoyl)piperazin-1-yl]prop-2-en-1-one
Synonyms:
1-[4-(prop-2-enoyl)piperazin-1-yl]prop-2-en-1-one
Molecular Weight (g/mol):
194.23
Molecular Formula:
C10H14N2O2
InChi Key:
YERHJBPPDGHCRJ-UHFFFAOYSA-N
SMILES:
C=CC(=O)N1CCN(CC1)C(=O)C=C