missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diacryloylpiperazine, MP Biomedicals

Product Code. 11411652 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 11411652

Brand: MP Biomedicals 04809239

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 6342-17-2
Molecular Formula C10H14N2O2
Quantity 10 g
Synonym 1,4-diacryloylpiperazine, diacrylylpiperazine, 1,4-bis acryloyl piperazine, 1,4-diacrylylpiperazine, 1,1'-piperazine-1,4-diyl bis prop-2-en-1-one, piperazine, 1,4-bis 1-oxo-2-propenyl, 1-4-prop-2-enoyl piperazin-1-yl prop-2-en-1-one, 1,1'-piperazine-1,4-diylbisprop-2-en-1-one, acmc-20ap2o, bicl800
InChI Key YERHJBPPDGHCRJ-UHFFFAOYSA-N
SMILES C=CC(=O)N1CCN(CC1)C(=O)C=C
IUPAC Name 1-(4-prop-2-enoylpiperazin-1-yl)prop-2-en-1-one
Molecular Weight (g/mol) 194.234
PubChem CID 193422
Chemical Name or Material 1, 4-Diacryloylpiperazine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.