CAS RN 6341-24-8
CAS RN 6341-24-8
IUPAC Name:
2-amino-3-(1H-imidazol-5-yl)propanoic acid hydrochloridyl
Synonyms:
histidine hydrochloridyl
Molecular Weight (g/mol):
190.61
Molecular Formula:
C6H9ClN3O2
InChi Key:
IRHBVTLJVRUHBJ-UHFFFAOYNA-N
SMILES:
[Cl].NC(CC1=CN=CN1)C(O)=O