missing translation for 'onlineSavingsMsg'
Learn More

D-histidine monohydrochloride hydrate, 99.2%, MP Biomedicals™

Product Code. 11304419 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 11304419

Brand: MP Biomedicals 0210194925

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 6341-24-8
Color White
Assay Percent Range 0.992
Molecular Formula C6H9ClN3O2
MDL Number MFCD00012702,MFCD00150081
Synonym d-histidine hydrochloride monohydrate, d-histidine monohydrochloride monohydrate, d-histidine hydrate hydrochloride, d-alpha-amino-beta-4-imidazolyl propionic acid monohydrochloride, d-histidine monohydrochloride monohydrate tlc, d-histidine monohydrochloride monohydrate at, d-2-degrees +/->>u-3-4-ssasso>>u +/-uea micro yennieani, r-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride monohydrate
InChI Key IRHBVTLJVRUHBJ-UHFFFAOYNA-N
SMILES [Cl].NC(CC1=CN=CN1)C(O)=O
IUPAC Name 2-amino-3-(1H-imidazol-5-yl)propanoic acid hydrochloridyl
Molecular Weight (g/mol) 190.61
Quantity 25 g
PubChem CID 15556510
Formula Weight 191.62
Physical Form Powder
Chemical Name or Material D-Histidine monohydrochloride hydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.