CAS RN 6308-22-1
CAS RN 6308-22-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
AKOS B018304, MedChemExpress
MedChemExpress AKOS B018304 is an arylalkylidene derivative, with polar substitution at para-position.