Learn More
AKOS B018304, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
AKOS B018304 is an arylalkylidene derivative, with polar substitution at para-position.
Specifications
Specifications
| Chemical Name or Material | AKOS B018304 |
| Physical Form | Solid |
| CAS | 6308-22-1 |
| Color | Gray |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C10H6N2O3S2 |
| Solubility Information | DMSO : ≥ 77.5 mg/mL (291.03 mM) |
| SMILES | O=C1NC(S/C1=C/C2=CC=CC=C2[N+]([O-])=O)=S |
| Molecular Weight (g/mol) | 266.3 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.