CAS RN 6304-27-4
CAS RN 6304-27-4
IUPAC Name:
dimethyl[2-(pyridin-2-yl)ethyl]amine
Synonyms:
dimethyl[2-(pyridin-2-yl)ethyl]amine
Molecular Weight (g/mol):
150.23
Molecular Formula:
C9H14N2
InChi Key:
FXHJGPDCPMCUKW-UHFFFAOYSA-N
SMILES:
CN(C)CCC1=CC=CC=N1